Please login to your online account to display your discounted pricing

anti-C-Reactive Protein Polyclonal, Thermo Scientific™

C-Reactive Protein Polyclonal antibody PA1-18303 from Thermo Scientific™ specifically detects C-Reactive Protein in Human samples, and it is validated for Western Blot

$506.05 - $506.05

Specifications

Antigen C-Reactive Protein
Cross Reactivity Human
Format Serum
Host Species Sheep
Immunogen Recombinant human C-Reactive protein.
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

01-670-557

 
pierce biotechnology inc
PA118304
Unlabeled 100μL Each for $506.05

01-670-556

 
pierce biotechnology inc
PA118303
Unlabeled 100μL Each for $506.05
Description & Specifications

Specifications

Antigen C-Reactive Protein
Cross Reactivity Human
Format Serum
Host Species Sheep
Immunogen Recombinant human C-Reactive protein.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Avoid Freeze/Thaw Cycles

C-Reactive Protein Polyclonal antibody PA1-18303 from Thermo Scientific™ specifically detects C-Reactive Protein in Human samples, and it is validated for Western Blot

C Reactive Protein is a major acute phase reactant synthesized primarily in the liver hepatocytes. It is a pentraxin (cyclic pentameric protein) compound of five identical nonglycosylated subunits of 206 amino acids each (m.w. 24 kDa), that are bound noncovalently to form the physiologic CRP molecule (m.w. 117.5 kDa). C Reactive Protein mediates activities associated with preimmune nonspecific host resistance. It is opsonic, an initiator of the classical complement cascade and an activator of monocytes/macrophages. CRP also binds to several nuclear components including chromatin, histones and snRNP, suggesting that it may play a role as a scavenger during cell necrosis.
Host Species: Goat
Cross Reactivity: Human
Isotype: IgG
Immunogen: Recombinant human C-Reactive protein.
Formulation: 100μL unpurified Antibody in whole serum, with no preservative;
Synonyms: CRP, PTX1
Uniprot Accession: P02741
Gene Symbol: CRP
Entrez GeneID: 1401
PA1-18303 detects C-Reactive Protein in Human samples. PA1-18303 has been successfully used in Western Blot procedures. PA1-18303 The immunogen is Recombinant human C-Reactive protein..