Please login to your online account to display your discounted pricing

anti-c-Myc, Polyclonal, Novus Biologicals™

$144.63 - $362.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to the human c-Myc protein (between residues 400-450) [UniProt P01106]
Antigen c-Myc
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB600336SS

 
novus biologicals canada
NB600336SS
Unlabeled 0.025mL Each for $144.63

NB600336

 
novus biologicals canada
NB600336
Unlabeled 0.1mL Each for $362.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to the human c-Myc protein (between residues 400-450) [UniProt P01106]
Antigen c-Myc
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Chromatin Immunoprecipitation
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blot
Test Specificity Non-species specific when used to detect the c-Myc tag. When used to detect the c-Myc protein this antibody reacts with human and mouse.
Gene Accession No. P01106
Gene ID (Entrez) 4609
Formulation Tris-glycine, 150mM NaCl
Concentration 1.37mg/mL