Please login to your online account to display your discounted pricing

anti-c-Myc, Polyclonal, Novus Biologicals™

$307.00 - $307.00

Specifications

Host Species Chicken
Isotype IgG
Immunogen Chickens were immunized with a synthetic peptide representing amino acid residues 410-419 (EQKLISEEDL) of human myc conjugated to KLH.
Antigen c-Myc
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB600334

 
novus biologicals canada
NB600334
Unlabeled 0.1mg Each for $307.00
Description & Specifications

Specifications

Host Species Chicken
Isotype IgG
Immunogen Chickens were immunized with a synthetic peptide representing amino acid residues 410-419 (EQKLISEEDL) of human myc conjugated to KLH.
Antigen c-Myc
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene Accession No. P01106
Gene ID (Entrez) 4609
Formulation phosphate Buffered Saline (PBS)
Concentration 1.0mg/mL