Please login to your online account to display your discounted pricing

anti-c-Myc, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB6003341MG

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB6003341MG

  • / Each
 

Description & Specifications

Specifications

Formulation phosphate Buffered Saline (PBS)
Gene ID (Entrez) 4609
Concentration 1.0mg/mL
Applications ELISA
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blotting
Conjugate Unlabeled
Gene Accession No. P01106
Immunogen Chickens were immunized with a synthetic peptide representing amino acid residues 410-419 (EQKLISEEDL) of human myc conjugated to KLH.
Host Species Chicken
Monoclonal or Polyclonal Polyclonal
Isotype IgG
Regulatory Status RUO
Quantity 1mg
Format Affinity Purified
Antigen c-Myc
Cross Reactivity Non-species specific
Primary or Secondary Primary