Please login to your online account to display your discounted pricing

anti-c-Maf, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224551

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224551

  • / Each
 

Description & Specifications

Specifications

Antigen c-Maf
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Affinity Purified
Formulation 100μg in 200μL PBS containing 0.05% BSA
Gene Accession No. O75444
Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 75-110 of human c-maf was used as the immunogen
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 4094
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, CRA_b, and CRA_c), 93% homologous in monkey, 87% homologous in chimpanzee and 81% homologous in cow and horse.
Target Species Bovine
Target Species Human
Target Species Murine
Target Species Rat