Please login to your online account to display your discounted pricing

anti-c-Maf, Polyclonal, Novus Biologicals™

$139.00 - $307.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 75-110 of human c-maf was used as the immunogen
Antigen c-Maf
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224551

 
novus biologicals canada
NBP224551
Unlabeled 0.1mg Each for $307.00

NBP224551SS

 
novus biologicals canada
NBP224551SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide corresponding to amino acids 75-110 of human c-maf was used as the immunogen
Antigen c-Maf
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human (isoforms CRA_a, CRA_b, and CRA_c), 93% homologous in monkey, 87% homologous in chimpanzee and 81% homologous in cow and horse.
Gene Accession No. O75444
Gene ID (Entrez) 4094
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL