Please login to your online account to display your discounted pricing

anti-c-Fos Polyclonal, Thermo Scientific™

c-Fos Polyclonal Ab PA1-18328 from Thermo Scientific™ specifically detects c-Fos in Hamster, Rb and Rt samples, and it is validated for IHC and WB

$543.75 - $578.55

Specifications

Antigen c-Fos
Cross Reactivity Hamster
Cross Reactivity Rabbit
Cross Reactivity Rat
Format Serum
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

01-670-581

 
pierce biotechnology inc
PA118328
Unlabeled 100μL Each for $543.75

01-670-583

 
pierce biotechnology inc
PA118330
Unlabeled 50μg Each for $543.75

01-670-582

 
pierce biotechnology inc
PA118329
Unlabeled 500μg Each for $578.55
Description & Specifications

Specifications

Antigen c-Fos
Cross Reactivity Hamster
Cross Reactivity Rabbit
Cross Reactivity Rat
Format Serum
Host Species Sheep
Immunogen Synthetic peptide conjugated to diphtheria toxoid residues M(2) F S G F N A D Y E A S S S R C(17).
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Avoid Freeze/Thaw Cycles

c-Fos Polyclonal Ab PA1-18328 from Thermo Scientific™ specifically detects c-Fos in Hamster, Rb and Rt samples, and it is validated for IHC and WB

an immediate early gene encoding a nuclear protein involved in signal transduction.
Host Species: Sheep
Cross Reactivity: Hamster, Rabbit, Rat
Isotype: IgG
Immunogen: Synthetic peptide conjugated to diphtheria toxoid residues M(2) F S G F N A D Y E A S S S R C(17).
Formulation: 100μL unpurified Antibody in whole serum, with no preservative;
Synonyms: Cellular Oncogene Fos
Uniprot Accession: P12841
Gene Symbol: Fos
Entrez GeneID: 100348962, 314322
PA1-18328 detects c-Fos in Hamster, Rabbit and Rat samples. PA1-18328 has been successfully used in Immunohistochemistry and Western Blot procedures. PA1-18328 The immunogen is Synthetic peptide conjugated to diphtheria toxoid residues M(2) F S G F N A D Y E A S S S R C(17)..