Please login to your online account to display your discounted pricing

anti-c-Abl, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11081723SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11081723T

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P00519
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Primary or Secondary Primary
Gene ID (Entrez) 25
Concentration 1mg/mL
Formulation Tris-citrate/phosphate, pH 7-8
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Human
Cross Reactivity Mouse
Immunogen Synthetic peptide made to an internal portion of human c Abl (within residues 700-800). [Swiss-Prot No. P00519]
Antigen c-Abl
Conjugate Unlabeled
Host Species Rabbit
Regulatory Status RUO
Quantity 0.025mL
Format Affinity Purified