Please login to your online account to display your discounted pricing

anti-c-Abl, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11081723

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11081723

  • / Each
 

Description & Specifications

Specifications

Antigen c-Abl
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Formulation Tris-citrate/phosphate, pH 7-8
Gene Accession No. P00519
Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human c Abl (within residues 700-800). [Swiss-Prot No. P00519]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 25
Target Species Human
Target Species Murine