Please login to your online account to display your discounted pricing

anti-c-Abl, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human c Abl (within residues 700-800). [Swiss-Prot No. P00519]
Antigen c-Abl
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11081723T

 
novus biologicals canada
NB11081723SS
Unlabeled 0.025mL Each for $109.00

NB11081723

 
novus biologicals canada
NB11081723
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic peptide made to an internal portion of human c Abl (within residues 700-800). [Swiss-Prot No. P00519]
Antigen c-Abl
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Gene Accession No. P00519
Gene ID (Entrez) 25
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL