Please login to your online account to display your discounted pricing

anti-BTLA/CD272, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224731

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224731

  • / Each
 

Description & Specifications

Specifications

Formulation 100μg in 200μL PBS containing 0.2% gelatin
Conjugate Unlabeled
Test Specificity The amino acid sequence used as immunogen is 100% homologous in mouse,88% homologous in rat, 80% homologous inchickenand 76% homologous in cow.
Host Species Rabbit
Concentration 0.5mg/mL
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 151888
Regulatory Status RUO
Cross Reactivity Mouse
Format Purified
Quantity 0.1mg
Isotype IgG
Antigen BTLA/CD272
Primary or Secondary Primary
Applications Western Blotting
Immunogen A portion of amino acids 1-50 of mouse BTLA was used as the immunogen.