Please login to your online account to display your discounted pricing

anti-BTLA/CD272, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 1-50 of mouse BTLA was used as the immunogen.
Antigen BTLA/CD272
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224731

 
novus biologicals canada
NBP224731
Unlabeled 0.1mg Each for $255.80

NBP224731SS

 
novus biologicals canada
NBP224731SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 1-50 of mouse BTLA was used as the immunogen.
Antigen BTLA/CD272
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in mouse,88% homologous in rat, 80% homologous inchickenand 76% homologous in cow.
Gene ID (Entrez) 151888
Formulation 100μg in 200μL PBS containing 0.2% gelatin
Concentration 0.5mg/mL