Please login to your online account to display your discounted pricing

anti-BTK (p Tyr223), Polyclonal, Novus Biologicals™

$139.00 - $384.00

Specifications

Host Species Rabbit
Immunogen A synthetic phosphorylated peptide made to a peptide surrounding amino acid 223 of the human BTK protein [UniProt Q06187]
Antigen BTK (p Tyr223)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP178295

 
novus biologicals canada
NBP178295
Unlabeled 0.1mL Each for $384.00

NBP178295SS

 
novus biologicals canada
NBP178295SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic phosphorylated peptide made to a peptide surrounding amino acid 223 of the human BTK protein [UniProt Q06187]
Antigen BTK (p Tyr223)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Gene Accession No. Q06187
Gene ID (Entrez) 695
Formulation PBS, 30% glycerol
Concentration 1.09mg/mL