Please login to your online account to display your discounted pricing

anti-BST2, Polyclonal, Novus Biologicals™

$139.00 - $257.41

Specifications

Host Species Rabbit
Immunogen ITTLNHkLQDASAEVERLR 120-138
Antigen BST2
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227349

 
novus biologicals canada
NBP227349
Unlabeled 0.1mg Each for $257.41

NBP227349SS

 
novus biologicals canada
NBP227349SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen ITTLNHkLQDASAEVERLR 120-138
Antigen BST2
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Gene Accession No. Q10589
Gene ID (Entrez) 684
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA.
Concentration 0.5mg/mL