Please login to your online account to display your discounted pricing

anti-BRIP1/FANCJ, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100416SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100416SS

  • / Each
 

Description & Specifications

Specifications

Quantity 0.025mL
Monoclonal or Polyclonal Polyclonal
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Conjugate Unlabeled
Gene Accession No. Q9BX63
Regulatory Status RUO
Gene ID (Entrez) 83990
Format Antiserum
Primary or Secondary Primary
Immunogen A synthetic peptide within the extreme C-terminus of human FANCJ. [Swiss-Prot No. Q9BX63]
Cross Reactivity Human
Cross Reactivity Mouse
Host Species Rabbit
Antigen BRIP1/FANCJ