Please login to your online account to display your discounted pricing

anti-BRIP1/FANCJ, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB100416

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB100416

  • / Each
 

Description & Specifications

Specifications

Host Species Rabbit
Gene ID (Entrez) 83990
Conjugate Unlabeled
Gene Accession No. Q9BX63
Cross Reactivity Human
Cross Reactivity Mouse
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blotting
Quantity 0.1mL
Antigen BRIP1/FANCJ
Monoclonal or Polyclonal Polyclonal
Format Antiserum
Immunogen A synthetic peptide within the extreme C-terminus of human FANCJ. [Swiss-Prot No. Q9BX63]
Primary or Secondary Primary
Regulatory Status RUO