Please login to your online account to display your discounted pricing

anti-BRIP1/FANCJ, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A synthetic peptide within the extreme C-terminus of human FANCJ. [Swiss-Prot No. Q9BX63]
Antigen BRIP1/FANCJ
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100416SS

 
novus biologicals canada
NB100416SS
Unlabeled 0.025mL Each for $139.00

NB100416

 
novus biologicals canada
NB100416
Unlabeled 0.1mL Each for $329.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide within the extreme C-terminus of human FANCJ. [Swiss-Prot No. Q9BX63]
Antigen BRIP1/FANCJ
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Gene Accession No. Q9BX63
Gene ID (Entrez) 83990