Please login to your online account to display your discounted pricing

anti-Bre1, Polyclonal, Novus Biologicals™

$590.00 - $590.00

Specifications

Host Species Rabbit
Immunogen In vivo generated recombinant protein fragment
Antigen Bre1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

40-280-002

 
novus biologicals canada
40280002
Unlabeled 0.05mg Each for $590.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen In vivo generated recombinant protein fragment
Antigen Bre1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Chromatin Immunoprecipitation
Applications ELISA
Applications Western Blotting
Test Specificity Drosophila.
Gene ID (Entrez) 38652
Formulation 20mM Potassium phosphate, 150mM Sodium Chloride, pH 7.0