Please login to your online account to display your discounted pricing

anti-BRD4, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056487SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056487T

  • / Each
 

Description & Specifications

Specifications

Antigen BRD4
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Primate
Cross Reactivity Rat
Format Purified
Formulation PBS containing 0.05% BSA
Gene Accession No. O60885
Host Species Rabbit
Immunogen A portion of amino acids 150-200 of human BRD4 was used as the immunogen (NP_490597).
Quantity 0.025mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 23476
Test Specificity The amino acid sequence used as immunogen has 100% identity in human, chimp, mouse and rat. BRD4 is expressed in two forms, the long form is approximately 1400 amino acids and the short form is approximately 722 amino acids. Human, Chimpanzee, Mouse and Rat. Not yet tested in other species.
Target Species Human
Target Species Murine
Target Species Primate
Target Species Rat