Please login to your online account to display your discounted pricing

anti-BRD4, Polyclonal, Novus Biologicals™

$139.00 - $329.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 150-200 of human BRD4 was used as the immunogen (NP_490597).
Antigen BRD4
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB10056487

 
novus biologicals canada
NB10056487
Unlabeled 0.1mg Each for $329.00

NB10056487T

 
novus biologicals canada
NB10056487SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 150-200 of human BRD4 was used as the immunogen (NP_490597).
Antigen BRD4
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen has 100% identity in human, chimp, mouse and rat. BRD4 is expressed in two forms, the long form is approximately 1400 amino acids and the short form is approximately 722 amino acids. Human, Chimpanzee, Mouse and Rat. Not yet tested in other species.
Gene Accession No. O60885
Gene ID (Entrez) 23476
Formulation PBS containing 0.05% BSA
Concentration 0.5mg/mL