Please login to your online account to display your discounted pricing

anti-Brachyury, Polyclonal, Novus Biologicals™

$109.00 - $307.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 350-400 of human Brachyury was used as the immunogen.
Antigen Brachyury
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224676

 
novus biologicals canada
NBP224676
Unlabeled 0.1mg Each for $307.00

NBP224676SS

 
novus biologicals canada
NBP224676SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 350-400 of human Brachyury was used as the immunogen.
Antigen Brachyury
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee, horse and cow, 93% homologous in mouse, rat, dog, monkey and 73% homologous in chicken.
Gene Accession No. O15178
Gene ID (Entrez) 6862
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL