Please login to your online account to display your discounted pricing

anti-BP1, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made specifically to the human BP1 protein sequence (between residues 1-60) [UniProt No. Q92988]. There is no homology with the peptide sequences used for immunogen of this antibody and the DLX4 or DLX7 proteins.
Antigen BP1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB100481

 
novus biologicals canada
NB100481
Unlabeled 0.1mL Each for $309.05

NB100481SS

 
novus biologicals canada
NB100481SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made specifically to the human BP1 protein sequence (between residues 1-60) [UniProt No. Q92988]. There is no homology with the peptide sequences used for immunogen of this antibody and the DLX4 or DLX7 proteins.
Antigen BP1
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Test Specificity This is specific for human BP1 (DLX4 variant 1). Berg PE, Kirolikar S. DLX4 (distal-less homeobox 4). Atlas Genet Cytogenet Oncol Haematol. January 2011. Human. Immunogen sequence has 82% homology with mouse.
Gene Accession No. Q92988
Gene ID (Entrez) 1748
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL