Please login to your online account to display your discounted pricing

anti-Bovine Serum Albumin, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224936

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224936

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Formulation 100μg in 200μL PBS containing 0.05% BSA
Antigen Bovine Serum Albumin
Gene ID (Entrez) 280717
Conjugate Unlabeled
Host Species Rabbit
Concentration 0.5mg/mL
Regulatory Status RUO
Format Purified
Immunogen BSA
Cross Reactivity Bovine
Applications Western Blotting
Monoclonal or Polyclonal Polyclonal
Quantity 0.1mg