Please login to your online account to display your discounted pricing

anti-Bovine Serum Albumin, Polyclonal, Novus Biologicals™

$109.00 - $329.00

Specifications

Host Species Rabbit
Immunogen BSA
Antigen Bovine Serum Albumin
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224936SS

 
novus biologicals canada
NBP224936SS
Unlabeled 0.025mg Each for $109.00

NBP224936

 
novus biologicals canada
NBP224936
Unlabeled 0.1mg Each for $329.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen BSA
Antigen Bovine Serum Albumin
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Gene ID (Entrez) 280717
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL