Please login to your online account to display your discounted pricing

anti-Bovine Serum Albumin, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224936SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224936SS

  • / Each
 

Description & Specifications

Specifications

Applications Western Blotting
Conjugate Unlabeled
Concentration 0.5mg/mL
Gene ID (Entrez) 280717
Formulation 100μg in 200μL PBS containing 0.05% BSA
Regulatory Status RUO
Immunogen BSA
Host Species Rabbit
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Bovine
Antigen Bovine Serum Albumin
Primary or Secondary Primary
Quantity 0.025mg
Format Purified