Please login to your online account to display your discounted pricing

anti-Bone marrow stromal cell antigen 1/CD157, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 30-80 of human BST1 was used as the immunogen for this antibody.
Antigen Bone marrow stromal cell antigen 1/CD157
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224581SS

 
novus biologicals canada
NBP224581SS
Unlabeled 0.025mg Each for $109.00

NBP224581

 
novus biologicals canada
NBP224581
Unlabeled 0.1mg Each for $255.80
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 30-80 of human BST1 was used as the immunogen for this antibody.
Antigen Bone marrow stromal cell antigen 1/CD157
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunohistochemistry (Paraffin)
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human isoforms CRA_a, CRA_b, CRA_e and CRA_f (ranging between 440 and 589 amino acids), monkey, chicken, horse, mouse and chimpanzee, 93% homologous in rat, 87% homologous in zebrafish and xenopus and 81% homologous in pufferfish.
Gene Accession No. Q10588
Gene ID (Entrez) 683
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL