Please login to your online account to display your discounted pricing

anti-Bone marrow stromal cell antigen 1/CD157, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224581SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224581SS

  • / Each
 

Description & Specifications

Specifications

Primary or Secondary Primary
Formulation 100μg antibody in 200μL PBS containing 0.05% BSA
Monoclonal or Polyclonal Polyclonal
Quantity 0.025mg
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human isoforms CRA_a, CRA_b, CRA_e and CRA_f (ranging between 440 and 589 amino acids), monkey, chicken, horse, mouse and chimpanzee, 93% homologous in rat, 87% homologous in zebrafish and xenopus and 81% homologous in pufferfish.
Gene ID (Entrez) 683
Concentration 0.5mg/mL
Cross Reactivity Human
Cross Reactivity Primate
Gene Accession No. Q10588
Host Species Rabbit
Applications Immunohistochemistry (Paraffin)
Antigen Bone marrow stromal cell antigen 1/CD157
Format Affinity Purified
Conjugate Unlabeled
Regulatory Status RUO
Immunogen A portion of amino acids 30-80 of human BST1 was used as the immunogen for this antibody.