Please login to your online account to display your discounted pricing

anti-BNIP3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP177683

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP177683

  • / Each
 

Description & Specifications

Specifications

Antigen BNIP3
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Concentration 1.17mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Mouse
Format Affinity Purified
Formulation PBS, 30% glycerol
Gene Accession No. Q12983
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human BNIP3 protein (within residues 50-150). [Swiss-Prot Q12983]
Quantity 0.1mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 664
Target Species Human
Target Species Murine