Please login to your online account to display your discounted pricing

anti-BNIP3, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide against an N-terminal region (within residues 1-50) of the human BNIP3 protein. [UniProt No. Q12983]
Antigen BNIP3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11060562

 
novus biologicals canada
NB11060562
Unlabeled 0.1mL Each for $309.05

NB11060562T

 
novus biologicals canada
NB11060562SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide against an N-terminal region (within residues 1-50) of the human BNIP3 protein. [UniProt No. Q12983]
Antigen BNIP3
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Test Specificity Human. Immunogen sequence has 93% homology to feline and 87% homology to bovine and canine proteins. Does not work in mouse.
Gene Accession No. Q12983
Gene ID (Entrez) 664
Formulation Tris-citrate/phosphate, pH 7-8
Concentration 1mg/mL