Please login to your online account to display your discounted pricing

anti-BNIP3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11060562SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11060562T

  • / Each
 

Description & Specifications

Specifications

Antigen BNIP3
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Concentration 1mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation Tris-citrate/phosphate, pH 7-8
Gene Accession No. Q12983
Host Species Rabbit
Immunogen A synthetic peptide against an N-terminal region (within residues 1-50) of the human BNIP3 protein. [UniProt No. Q12983]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 664
Test Specificity Human. Immunogen sequence has 93% homology to feline and 87% homology to bovine and canine proteins. Does not work in mouse.
Target Species Human