Please login to your online account to display your discounted pricing

anti-BNIP3, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB11060562

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB11060562

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. Q12983
Gene ID (Entrez) 664
Formulation Tris-citrate/phosphate, pH 7-8
Test Specificity Human. Immunogen sequence has 93% homology to feline and 87% homology to bovine and canine proteins. Does not work in mouse.
Antigen BNIP3
Quantity 0.1mL
Concentration 1mg/mL
Conjugate Unlabeled
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blotting
Format Affinity Purified
Host Species Rabbit
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Cross Reactivity Human
Immunogen A synthetic peptide against an N-terminal region (within residues 1-50) of the human BNIP3 protein. [UniProt No. Q12983]