Please login to your online account to display your discounted pricing

anti-BNC1, Polyclonal, Novus Biologicals™

$139.00 - $259.02

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 330-380 of human Bnc 1 protein was used as the immunogen.
Antigen BNC1
Monoclonal or Polyclonal Polyclonal
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224721

 
novus biologicals canada
NBP224721
Unlabeled 0.1mg Each for $259.02

NBP224721SS

 
novus biologicals canada
NBP224721SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Isotype IgG
Immunogen A portion of amino acids 330-380 of human Bnc 1 protein was used as the immunogen.
Antigen BNC1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, monkey and dog, 91% homologous in horse, mouse and rat, and 83% homologous in cow and opossum.
Gene Accession No. Q01954
Gene ID (Entrez) 646
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL