Please login to your online account to display your discounted pricing

anti-BMI-1, Polyclonal, Novus Biologicals™

$139.00 - $307.00

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 270-320 of human BMI1 was used as the immunogen.
Antigen BMI-1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224693

 
novus biologicals canada
NBP224693
Unlabeled 0.1mg Each for $307.00

NBP224693SS

 
novus biologicals canada
NBP224693SS
Unlabeled 0.025mg Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 270-320 of human BMI1 was used as the immunogen.
Antigen BMI-1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry (Paraffin)
Applications Western Blot
Test Specificity The amino acid sequence used is 100% homologous in human (isoforms 1 and 2), mouse, rat, dog and bovine.
Gene Accession No. P35226
Gene ID (Entrez) 648
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL