Please login to your online account to display your discounted pricing

anti-Blooms Syndrome Protein Blm (p Thr99), Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP146851SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP146851SS

  • / Each
 

Description & Specifications

Specifications

Antigen Blooms Syndrome Protein Blm (p Thr99)
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Concentration 4.8mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Format Affinity Purified
Formulation 0.2M glycine, 1M Tris-HCl, pH 8.5
Gene Accession No. P54132
Host Species Rabbit
Immunogen Synthetic phospho-peptide surrounding the Blooms Syndrome Protein Blm Threonine 99 region [UniProt No. P54132]
Quantity 0.025mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 641
Target Species Human