Please login to your online account to display your discounted pricing

anti-Blooms Syndrome Protein Blm (p Thr99), Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Synthetic phospho-peptide surrounding the Blooms Syndrome Protein Blm Threonine 99 region [UniProt No. P54132]
Antigen Blooms Syndrome Protein Blm (p Thr99)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP146851SS

 
novus biologicals canada
NBP146851SS
Unlabeled 0.025mL Each for $139.00

NBP146851

 
novus biologicals canada
NBP146851
Unlabeled 0.05mL Each for $309.05
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Synthetic phospho-peptide surrounding the Blooms Syndrome Protein Blm Threonine 99 region [UniProt No. P54132]
Antigen Blooms Syndrome Protein Blm (p Thr99)
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Gene Accession No. P54132
Gene ID (Entrez) 641
Formulation 0.2M glycine, 1M Tris-HCl, pH 8.5
Concentration 4.8mg/mL