Please login to your online account to display your discounted pricing

anti-BLIMP1/PRDM1, Polyclonal, Novus Biologicals™

$109.00 - $255.80

Specifications

Host Species Rabbit
Immunogen Amino acids 692-719 of human BLIMP-1 (PRDM1) were used as the immunogen for this antibody.
Antigen BLIMP1/PRDM1
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP227180

 
novus biologicals canada
NBP227180
Unlabeled 0.1mL Each for $255.80

NBP227180SS

 
novus biologicals canada
NBP227180SS
Unlabeled 0.025mL Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Amino acids 692-719 of human BLIMP-1 (PRDM1) were used as the immunogen for this antibody.
Antigen BLIMP1/PRDM1
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blot
Test Specificity It is 100% homologous in isoforms 1 and 2 of human BLIMP-1 (PRDM1).
Gene ID (Entrez) 639
Formulation 100μL; neat rabbit sera (antibody) with 50% glycerol.