Please login to your online account to display your discounted pricing

anti-BID, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056106

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056106

  • / Each
 

Description & Specifications

Specifications

Antigen BID
Applications Immunohistochemistry
Applications Immunohistochemistry (Frozen)
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Western Blot
Conjugate Unlabeled
Cross Reactivity Canine
Cross Reactivity Gerbil
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Format Antisera
Formulation Neat serum
Host Species Rabbit
Immunogen Full-length recombinant mouse Bid protein was used as immunogen.
Quantity 0.05mL
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 637
Test Specificity Full-length Bid is known to undergo cleavage/truncation (reviewed in Yin, 2006). Bid was initially found to be cleaved and activated by capspase-8 following death receptor activation. The term “Bid” was first used to described the caspase-8 cleaved/truncated C-terminal Bid. Bid can be also cleaved by other proteases such as Granzyme B, calpains and cathepsin. The C-terminal portion is considered to be the active Bid moiety. This active form can translocate from the cytosol to the mitochondria. Therefore, the appearance of Bid in the mitochondria is considered to be an indication of active Bid. However, it should also be noted that Bid has been shown to translocate to the mitochondria without cleavage in some model systems. The proteolytic cleavage of Bid usually occurs in the unstructured loop region between the alpha 2 and alpha 3 helices, which is between amino acids (aa) 41 and 79 of Bid. For example, the caspase-8/3 cleavage site is at 60 (human) and 59 (mouse). It should be noted that after cleavage, the smaller N-terminal portion of Bid is not necessarily separated from the larger C-terminal portion. The exact size of the C-terminal cleavage products depends on the Bid cleavage sites; generally 11-15 kDa C-truncated cleavage forms are generated. Shorter C-terminal Bid cleavage products may result from protease cleavage that involves both caspases and other proteases. Full length Bid is quite stable, but cleaved/truncated forms may have a short half-life (less than 1.5 h) and degrade rapidly. Therefore, a time course may be useful when detecting Bid cleavage fragments.
Target Species Canine
Target Species Gerbil
Target Species Human
Target Species Murine
Target Species Rat