Please login to your online account to display your discounted pricing

anti-Nerve Growth Factor beta Polyclonal, Thermo Scientific™

Nerve Growth Factor beta Polyclonal Ab PA1-18378 detects Nerve Growth Factor beta in Avian, Hu, Ms and Rt samples, and is validated for ELISA, IHC, Inhibition Assays and WB

$448.05 - $543.75

Specifications

Antigen Nerve Growth Factor beta
Cross Reactivity Avian
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

01-670-632

 
pierce biotechnology inc
PA118380
Unlabeled 50μg Each for $543.75

01-670-630

 
pierce biotechnology inc
PA118378
Unlabeled 20μg Each for $448.05
Description & Specifications

Specifications

Antigen Nerve Growth Factor beta
Cross Reactivity Avian
Cross Reactivity Chicken
Cross Reactivity Human
Cross Reactivity Mouse
Format Purified
Host Species Sheep
Immunogen Native mouse beta NGF purified from submaxillary salivary gland.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Avoid Freeze/Thaw Cycles

Nerve Growth Factor beta Polyclonal Ab PA1-18378 detects Nerve Growth Factor beta in Avian, Hu, Ms and Rt samples, and is validated for ELISA, IHC, Inhibition Assays and WB

Beta NGF is a nerve growth factor that is important for the development and maintenance of the sympathetic and sensory nervous systems. It stimulates division and differentiation of sympathetic and embryonic sensory neurons.
Host Species: Rabbit
Cross Reactivity: Avian, Human, Mouse, Rat
Isotype: IgG
Immunogen: Native mouse beta NGF purified from submaxillary salivary gland.
Formulation: 20μg purified Antibody (1mg/mL) no preservative;
Synonyms: beta Nerve Growth Factor
Uniprot Accession: P01138, P01139, P25427
Gene Symbol: NGF
Entrez GeneID: 18049, 310738, 4803
PA1-18378 detects Nerve Growth Factor beta in Avian, Human, Mouse and Rat samples. PA1-18378 has been successfully used in ELISA, Immunohistochemistry, Inhibition Assays and Western Blot procedures. PA1-18378 The immunogen is Native mouse beta NGF purified from submaxillary salivary gland..