Please login to your online account to display your discounted pricing

beta-Catenin Mouse anti-Human, Clone: 196618, R&D Systems

Mouse Monoclonal Antibody

$202.89 - $326.67

Specifications

Antigen beta-Catenin
Clone 196618
Cross Reactivity Human
Format Purified
Host Species Mouse
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

MAB13291050

 
r & d systems
RDSMAB13291050
Unlabeled 50μg Each for $202.89

MAB13291100

 
r & d systems
RDSMAB13291100
Unlabeled 100μg Each for $326.67
Description & Specifications

Specifications

Antigen beta-Catenin
Clone 196618
Cross Reactivity Human
Format Purified
Host Species Mouse
Regulatory Status RUO
Immunogen E.Coli-derived recombinant human β-Catenin. aa 2-781
Isotype IgG3
Monoclonal or Polyclonal Monoclonal
Primary or Secondary Primary
Purification Method Protein A or G purified from hybridoma culture supernatant

beta-Catenin Monoclonal antibody MAB13291 Specifically detects beta-Catenin in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blotting