Please login to your online account to display your discounted pricing

anti-beta-Arrestin 1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224626SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224626SS

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 408
Conjugate Unlabeled
Formulation 100μg in 200μL PBS containing 0.05% BSA
Applications Western Blotting
Primary or Secondary Primary
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, dog and bovine.
Concentration 0.5mg/mL
Monoclonal or Polyclonal Polyclonal
Format Purified
Regulatory Status RUO
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Zebrafish
Quantity 0.025mg
Host Species Rabbit
Antigen beta-Arrestin 1
Immunogen A portion of amino acids 320-370 of human Beta-arrestin 1 was used as the immunogen.