Please login to your online account to display your discounted pricing

anti-beta-Arrestin 1, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NBP224626

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NBP224626

  • / Each
 

Description & Specifications

Specifications

Antigen beta-Arrestin 1
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Bovine
Cross Reactivity Human
Cross Reactivity Mouse
Cross Reactivity Rat
Cross Reactivity Zebrafish
Format Purified
Formulation 100μg in 200μL PBS containing 0.05% BSA
Host Species Rabbit
Immunogen A portion of amino acids 320-370 of human Beta-arrestin 1 was used as the immunogen.
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 408
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, dog and bovine.
Target Species Bovine
Target Species Human
Target Species Murine
Target Species Rat
Target Species Zebrafish