Please login to your online account to display your discounted pricing

anti-beta-Arrestin 1, Polyclonal, Novus Biologicals™

$109.00 - $257.41

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 320-370 of human Beta-arrestin 1 was used as the immunogen.
Antigen beta-Arrestin 1
Monoclonal or Polyclonal Polyclonal
Format Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NBP224626

 
novus biologicals canada
NBP224626
Unlabeled 0.1mg Each for $257.41

NBP224626SS

 
novus biologicals canada
NBP224626SS
Unlabeled 0.025mg Each for $109.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A portion of amino acids 320-370 of human Beta-arrestin 1 was used as the immunogen.
Antigen beta-Arrestin 1
Monoclonal or Polyclonal Polyclonal
Format Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Test Specificity The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, dog and bovine.
Gene ID (Entrez) 408
Formulation 100μg in 200μL PBS containing 0.05% BSA
Concentration 0.5mg/mL