Please login to your online account to display your discounted pricing

anti-beta-Actin, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB10056874

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB10056874

  • / Each
 

Description & Specifications

Specifications

Antigen beta-Actin
Applications Immunohistochemistry (Paraffin)
Applications Simple Western
Applications Western Blot
Concentration 0.5mg/mL
Conjugate Unlabeled
Cross Reactivity Human
Cross Reactivity Invertebrate
Cross Reactivity Mouse
Cross Reactivity Rat
Format Purified
Formulation PBS containing 0.05% BSA
Host Species Rabbit
Immunogen Amino acids 2-16 (CDDDIAALVIDNGSG) of actin protein were used as the immunogen.
Quantity 0.1mg
Regulatory Status RUO
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
Gene ID (Entrez) 60
Test Specificity The NCBI accession number for actin is AAA51567. This can not be used to test heart or muscle lysate loading. Cross reacts with Human, Mouse and Rat. Invertebrate reactivity reported in scientific literature (PMID: 25240497).
Target Species Human
Target Species Invertebrate
Target Species Murine
Target Species Rat