Please login to your online account to display your discounted pricing

anti-beta-Actin, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB600532SS

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB600532SS

  • / Each
 

Description & Specifications

Specifications

Regulatory Status RUO
Gene Accession No. P60709
Concentration 1mg/mL
Cross Reactivity Human
Cross Reactivity Mouse
Formulation Tris-citrate/phosphate, pH 7-8
Gene ID (Entrez) 60
Applications Simple Western
Applications Western Blotting
Host Species Rabbit
Immunogen A region corresponding to the N-terminus of human Beta Actin [UniProt No. P60709]
Antigen beta-Actin
Quantity 0.025mL
Conjugate Unlabeled
Format Affinity Purified
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal