Please login to your online account to display your discounted pricing

anti-beta-Actin, Polyclonal, Novus Biologicals™


Manufacturer: novus biologicals canada  NB600532

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. NB600532

  • / Each
 

Description & Specifications

Specifications

Gene Accession No. P60709
Concentration 1mg/mL
Gene ID (Entrez) 60
Conjugate Unlabeled
Immunogen A region corresponding to the N-terminus of human Beta Actin [UniProt No. P60709]
Host Species Rabbit
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Cross Reactivity Human
Cross Reactivity Mouse
Formulation Tris-citrate/phosphate, pH 7-8
Applications Simple Western
Applications Western Blotting
Quantity 0.1mL
Regulatory Status RUO
Format Affinity Purified
Antigen beta-Actin