Please login to your online account to display your discounted pricing

anti-Beclin 2, Polyclonal, Novus Biologicals™

$109.00 - $309.05

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal region (within residues 1-100) of the human Beclin 2 protein. [Uniprot No. A8MW95]
Antigen Beclin 2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB11060982T

 
novus biologicals canada
NB11060982SS
Unlabeled 0.025mL Each for $109.00

NB11060982

 
novus biologicals canada
NB11060982
Unlabeled 0.1mL Each for $309.05
Description & Specifications

Specifications

Host Species Rabbit
Immunogen A synthetic peptide made to an N-terminal region (within residues 1-100) of the human Beclin 2 protein. [Uniprot No. A8MW95]
Antigen Beclin 2
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Western Blotting
Gene Accession No. A8MW95
Gene ID (Entrez) 441925
Formulation PBS, 30% glycerol
Concentration 0.9mg/mL