Please login to your online account to display your discounted pricing

anti-Beclin 1/ATG6, Polyclonal, Novus Biologicals™

$139.00 - $309.05

Specifications

Host Species Rabbit
Immunogen Full length human Beclin 1 protein [Swiss-Prot No. Q14457]
Antigen Beclin 1/ATG6
Monoclonal or Polyclonal Polyclonal
Format Antisera
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB500266

 
novus biologicals canada
NB500266
Unlabeled 0.1mL Each for $309.05

NB500266SS

 
novus biologicals canada
NB500266SS
Unlabeled 0.025mL Each for $139.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Full length human Beclin 1 protein [Swiss-Prot No. Q14457]
Antigen Beclin 1/ATG6
Monoclonal or Polyclonal Polyclonal
Format Antisera
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Western Blot
Gene Accession No. Q14457
Gene ID (Entrez) 8678