Please login to your online account to display your discounted pricing

anti-Beclin 1/ATG6, Polyclonal, Novus Biologicals™

$164.00 - $384.00

Specifications

Host Species Rabbit
Immunogen Internal synthetic peptide to human Beclin 1
Antigen Beclin 1/ATG6
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

NB500249SS

 
novus biologicals canada
NB500249SS
Unlabeled 0.025mL Each for $164.00

NB500249

 
novus biologicals canada
NB500249
Unlabeled 0.1mL Each for $384.00
Description & Specifications

Specifications

Host Species Rabbit
Immunogen Internal synthetic peptide to human Beclin 1
Antigen Beclin 1/ATG6
Monoclonal or Polyclonal Polyclonal
Format Affinity Purified
Primary or Secondary Primary
Regulatory Status RUO
Applications Immunocytochemistry
Applications Immunofluorescence
Applications Immunohistochemistry
Applications Immunohistochemistry (Paraffin)
Applications Immunoprecipitation
Applications Simple Western
Applications Western Blot
Gene Accession No. Q14457
Gene ID (Entrez) 8678
Formulation PBS
Concentration 1.0mg/mL