Please login to your online account to display your discounted pricing

anti-Beclin-1 Polyclonal, Thermo Scientific™

Beclin-1 Polyclonal antibody PA1-18287 from Thermo Scientific™ specifically detects Beclin-1 in Human samples, and it is validated for Immunofluorescence and Immunohistochemistry

$543.75 - $543.75

Specifications

Antigen Beclin-1
Cross Reactivity Human
Format Serum
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the C-terminal residues of human Beclin-1.
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

01-670-540

 
pierce biotechnology inc
PA118287
Unlabeled 100μL Each for $543.75
Description & Specifications

Specifications

Antigen Beclin-1
Cross Reactivity Human
Format Serum
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the C-terminal residues of human Beclin-1.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Avoid Freeze/Thaw Cycles

Beclin-1 Polyclonal antibody PA1-18287 from Thermo Scientific™ specifically detects Beclin-1 in Human samples, and it is validated for Immunofluorescence and Immunohistochemistry

Beclin-1 may play a role in antiviral host defense, as it protects against infection by a neurovirulent strain of Sindbis virus. Also, Beclin-1 may play a role in autophagy.
Host Species: Rabbit
Cross Reactivity: Human
Isotype: IgG
Immunogen: Synthetic peptide corresponding to the C-terminal residues of human Beclin-1.
Formulation: 100μL unpurified Antibody in whole serum, with no preservative;
Synonyms: BECN1, APG6, Coiled-coil myosin-like BCL2-interacting protein, Protein GT197
Uniprot Accession: Q14457
Gene Symbol: BECN1
Entrez GeneID: 8678
PA1-18287 detects Beclin-1 in Human samples. PA1-18287 has been successfully used in Immunofluorescence and Immunohistochemistry procedures. PA1-18287 The immunogen is Synthetic peptide corresponding to the C-terminal residues of human Beclin-1..

Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly