Please login to your online account to display your discounted pricing

anti-BDNF Polyclonal, Thermo Scientific™

BDNF Polyclonal Ab PA1-18357 from Thermo Scientific™ specifically detects BDNF in Hu, Ms and Rt samples, and it is validated for ELISA, IHC, Inhibition Assays and WB

$424.20 - $522.90

Specifications

Antigen BDNF
Cross Reactivity Human
Format Purified
Host Species Rabbit
Immunogen Recombinant human BDNF.
View More Specs
Products
Catalog Number Mfr. No. Conjugate Quantity Price Quantity    

01-670-609

 
pierce biotechnology inc
PA118357
Unlabeled 100μL Each for $522.90

01-670-615

 
pierce biotechnology inc
PA118363
Unlabeled 500μg Each for $480.90

01-670-621

 
pierce biotechnology inc
PA118369
Unlabeled 500μg Each for $494.55

01-670-622

 
pierce biotechnology inc
PA118370
Unlabeled 20μg Each for $424.20

01-670-627

 
pierce biotechnology inc
PA118375
Unlabeled 100μL Each for $522.90
Description & Specifications

Specifications

Antigen BDNF
Cross Reactivity Human
Format Purified
Host Species Rabbit
Immunogen Recombinant human BDNF.
Isotype IgG
Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Storage Requirements Avoid Freeze/Thaw Cycles

BDNF Polyclonal Ab PA1-18357 from Thermo Scientific™ specifically detects BDNF in Hu, Ms and Rt samples, and it is validated for ELISA, IHC, Inhibition Assays and WB

The protein encoded by this gene is a member of the nerve growth factor family. It is induced by cortical neurons, and is necessary for survival of striatal neurons in the brain. Expression of this gene is reduced in both Alzheimer's and Huntington disease patients. This gene may play a role in the regulation of stress response and in the biology of mood disorders. Multiple transcript variants encoding distinct isoforms have been described for this gene.
Host Species: Rabbit
Cross Reactivity: Human, Mouse, Rat
Isotype: IgG
Immunogen: Recombinant human BDNF.
Formulation: 100μL unpurified Antibody in whole serum, with no preservative;
Synonyms: Brain-derived neurotrophic factor, Abrineurin
Uniprot Accession: P21237, P23363, P23560
Gene Symbol: BDNF
Entrez GeneID: 12064, 24225, 627
PA1-18357 detects BDNF in Human, Mouse and Rat samples. PA1-18357 has been successfully used in ELISA, Immunohistochemistry, Inhibition Assays and Western Blot procedures. PA1-18357 The immunogen is Recombinant human BDNF..